2-Chlorophenylboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C1705-1G
C1705-5G
TCIAC1705-1GEA
26.4
EUR
TCIAC1705-1G
TCIAC1705-5G
2-Chlorophenylboronic acid (contains varying amounts of Anhydride)
2-Chlorophenylboronic acid
Formule:
C₆H₆BClO₂ |
MDL Nummer:
MFCD00674012 CAS nummer: 3900-89-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...