1,3,5,7-Cyclooctatetraene ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
Stabilised with hydroquinone (HQ)
C0505-1ML
C0505-5ML
TCIAC0505-1MLEA
41.8
EUR
TCIAC0505-1ML
TCIAC0505-5ML
1,3,5,7-Cyclooctatetraene ≥98.0% (by GC) stabilised
1,3,5,7-Cyclooctatetraene
Formule:
C₈H₈ Kookpunt: 143°C Smeltpunt: –7 °C Dichtheid: 0,925 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00004161 CAS nummer: 629-20-9 EINECS: 211-080-3 UN: 2358 ADR: 3,II |
|
Specification Test Results
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...