Phthalaldehyde ≥99.0% (by GC) for HPLC labeling
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
P0280-100G P0280-25G P0280-5G
TCIAP0280-100GEA 333 EUR
TCIAP0280-100G TCIAP0280-25G TCIAP0280-5G
Phthalaldehyde ≥99.0% (by GC) for HPLC labeling
Phthalaldehyde
Formule: C₈H₆O₂
MW: 134.13 g/mol
Kookpunt: 84°C
Vlampunt: 132
MDL Nummer: MFCD00003335
CAS nummer: 643-79-8
EINECS: 211-402-2
UN: 2811
ADR: 6.1 III
Merck Index: 14,07368
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4363edc3c76dd8d8d563ba70bd86f7102f83c951&vid=616d154019da07ffbee742061fa3b36b2d91169d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 55.0 to 58.0 deg-C
Effect test of HPLC derivatization to pass test