Tetramethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis) reducing reagent

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0852-25G T0852-5G
TCIAT0852-25GEA 88 EUR
TCIAT0852-25G TCIAT0852-5G
Tetramethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis) reducing reagent
Tetramethylammonium tetrahydroborate
Formule: C₄H₁₆BN
MDL Nummer: MFCD00011778
CAS nummer: 16883-45-7
EINECS: 240-917-5
UN: 3131
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b947ebd6c161e9aa250ef4dd8f4171384474d9a5&vid=9c5c6f73854d00a6a724ef6d62ccc2be8fe26b9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Sodium hypochlorite Method) min. 90.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR