3-(Methylthio)-1-propanol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0735-25G M0735-5G
TCIAM0735-25GEA 94.6 EUR
TCIAM0735-25G TCIAM0735-5G
3-(Methylthio)-1-propanol ≥99.0% (by GC)
3-(Methylthio)-1-propanol
Formule: C₄H₁₀OS
Kookpunt: 136°C
Dichtheid: 1,03 g/cm³ (20 °C)
Vlampunt: 87°C
MDL Nummer: MFCD00036560
CAS nummer: 505-10-2
EINECS: 208-004-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=032d0f6bb60c6800011721c13408500f6bd65f4a&vid=032d0f6bb60c6800011721c13408500f6bd65f4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR