2-(2-Chloroethoxy)ethanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C0169-25ML
C0169-500ML
C0169-100ML
TCIAC0169-25MLEA
19.8
EUR
TCIAC0169-25ML
TCIAC0169-500ML
TCIAC0169-100ML
2-(2-Chloroethoxy)ethanol ≥98.0% (by GC)
2-(2-Chloroethoxy)ethanol
Formule:
C₄H₉ClO₂ MW: 124.57 g/mol Kookpunt: 185°C Vlampunt: 107°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00002870 CAS nummer: 628-89-7 EINECS: 211-059-9 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...