1-Iodooctane ≥97.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with copper chip

I0321-25G I0321-500G I0321-100G
TCIAI0321-25GEA 18.7 EUR
TCIAI0321-25G TCIAI0321-500G TCIAI0321-100G
1-Iodooctane ≥97.0% (by GC) stabilised
1-Iodooctane
Formule: C₈H₁₇I
Kookpunt: 226°C
Smeltpunt: –46 °C
Dichtheid: 1,32 g/cm³ (20 °C)
Vlampunt: 87°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001105
CAS nummer: 629-27-6
EINECS: 211-084-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79df6f7fd083658f8d32df0a89aaa07d545ce021&vid=795dd93cde09225b2efab436c9f8251124a7a274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR