Benzylbromide ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: alpha-Bromotoluene
Stabilisatie: Stabilised with propylene oxide
B0411-100G B0411-25G B0411-500G
TCIAB0411-100GEA 30.8 EUR
TCIAB0411-100G TCIAB0411-25G TCIAB0411-500G
Benzylbromide ≥98.0% (by GC) stabilised
Benzylbromide
Formule: (C₆H₅)CH₂Br
Kookpunt: 199 °C (1013 hPa)
Smeltpunt: –4 °C
Dichtheid: 1,438 g/cm³ (20 °C)
Vlampunt: 79°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000172
CAS nummer: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1,II
Merck Index: 14,01128
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=420f7780e91b6f65b9365d6f828e2b0c2e524c29&vid=1a0754abc42af01a2d711e2948800a8a3933e3bc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %