Octanophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

O0149-25ML O0149-500ML O0149-100ML
TCIAO0149-25MLEA 47.3 EUR
TCIAO0149-25ML TCIAO0149-500ML TCIAO0149-100ML
Octanophenone ≥98.0% (by GC)
Octanophenone
Formule: C₆H₅C(O)(CH₂)₆CH₃
Kookpunt: 290°C
Smeltpunt: 22 °C
Dichtheid: 0,9434 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003554
CAS nummer: 1674-37-9
EINECS: 216-817-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ab943b2dd985544f00861918259616b388e4c48&vid=cb62018b9d8858070f9b0a86768afc6e13f18538&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR