Clioquinol ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C0187-25G
C0187-250G
TCIAC0187-25GEA
26.4
EUR
TCIAC0187-25G
TCIAC0187-250G
Clioquinol ≥97.0% (by titrimetric analysis)
Clioquinol
Formule:
C₉H₅ClINO Kookpunt: 365 °C (1013 hPa) Smeltpunt: 183…184 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006787 CAS nummer: 130-26-7 EINECS: 204-984-4 ADR: 6.1,III Merck Index: 14,05031 |
|
Specification Test Results
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...