Cloxiquina

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PROOMM0312.01EA 0 EUR
PROOMM0312.01
Cloxiquina
Cloxiquina
Formule: C₉H₆ClNO
MW: 179,61 g/mol
Kookpunt: 349 °C (1013 hPa)
Smeltpunt: 125 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006788
CAS nummer: 130-16-5
EINECS: 204-978-1
Merck Index: 12,02483

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR