L-(-)-tert-Leucine (H-Tle-OH) ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0147-1G
L0147-25G
L0147-5G
TCIAL0147-1GEA
17.6
EUR
TCIAL0147-1G
TCIAL0147-25G
TCIAL0147-5G
L-(-)-tert-Leucine (H-Tle-OH) ≥98.0% (by titrimetric analysis)
L-(-)-tert-Leucine (H-Tle-OH)
Formule:
C₆H₁₃NO₂ |
MDL Nummer:
MFCD00064218 CAS nummer: 20859-02-3 EINECS: 424-750-7 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Optical purity (LC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -8.0 to -10.0 deg (C=3, H2O) |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...