Perfluorobutane-1-sulphonyl fluoride ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride
P1098-25G P1098-250G
TCIAP1098-25GEA 55 EUR
TCIAP1098-25G TCIAP1098-250G
Perfluorobutane-1-sulphonyl fluoride ≥96.0% (by GC)
Perfluorobutane-1-sulphonyl fluoride
Formule: C₄F₁₀O₂S
MW: 302.09 g/mol
Kookpunt: 65°C
MDL Nummer: MFCD00007422
CAS nummer: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cff945bf5f96ee8ad809c2b92c28a0e8e5af59c&vid=7cff945bf5f96ee8ad809c2b92c28a0e8e5af59c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %