Isoluminol (4-aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis) chemiluminescence reagent
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-Amino-2,3-dihydro-1,4-phthalazinedione, 4-Aminophthaloylhydrazine, 6-Amino-2,3-dihydrophthalazine-1,4-dione, Isoluminol, 4-Aminophthalhydrazide
A5300-1G
A5300-10G
TCIAA5300-1GEA
70.4
EUR
TCIAA5300-1G
TCIAA5300-10G
Isoluminol (4-aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis) chemiluminescence reagent
Isoluminol (4-aminophthalhydrazide)
Formule:
C₈H₇N₃O₂ MW: 177.16 g/mol |
MDL Nummer:
MFCD00010560 CAS nummer: 3682-14-2 |
|
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to lump |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in Na2CO3 | almost transparency |
Water | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...