Octyltrimethylammonium chloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

O0178-25G O0178-100G O0178-5G
TCIAO0178-25GEA 84.7 EUR
TCIAO0178-25G TCIAO0178-100G TCIAO0178-5G
Octyltrimethylammonium chloride ≥98.0% (by titrimetric analysis)
Octyltrimethylammonium chloride
Formule: C₁₁H₂₆ClN
MDL Nummer: MFCD00059977
CAS nummer: 10108-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ef15817b20c177d7e0750a9b710589cd94b9bca&vid=984cabbc89762fb34626d8eb6510f1dd002b6896&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR