DL-tert-Leucine (H-DL-Tle-OH)

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-DL-Tle-OH

4005657.0001 4005657.0005
BACE4005657.0001EA 0 EUR
BACE4005657.0001 BACE4005657.0005
DL-tert-Leucine (H-DL-Tle-OH)
DL-tert-Leucine (H-DL-Tle-OH)
Formule: C₆H₁₃NO₂
MDL Nummer: MFCD00065933
CAS nummer: 33105-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR