2,5-Dimethylterephthalic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2208-25G D2208-5G
TCIAD2208-25GEA 1170 EUR
TCIAD2208-25G TCIAD2208-5G
2,5-Dimethylterephthalic acid ≥97.0% (by GC, titration analysis)
2,5-Dimethylterephthalic acid
Formule: C₁₀H₁₀O₄
MDL Nummer: MFCD00020292
CAS nummer: 6051-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f97971845cb9a4b0a98248c3669f4a457506f839&vid=f97971845cb9a4b0a98248c3669f4a457506f839&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR