4-Fluoro-DL-phenylalanine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4000519.0005 4000519.0025
BACE4000519.0005EA 0 EUR
BACE4000519.0005 BACE4000519.0025
4-Fluoro-DL-phenylalanine
4-Fluoro-DL-phenylalanine
Formule: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Nummer: MFCD00002600
CAS nummer: 51-65-0
EINECS: 200-113-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR