3-Fluoro-D-phenylalanine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-D-Phe(3-F)-OH

4025831.0001 4025831.0005
BACE4025831.0001EA 0 EUR
BACE4025831.0001 BACE4025831.0005
3-Fluoro-D-phenylalanine
3-Fluoro-D-phenylalanine
Formule: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Nummer: MFCD00066449
CAS nummer: 110117-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR