4-Fluoro-D-phenylalanine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4011120.0001 4011120.0005
BACE4011120.0001EA 0 EUR
BACE4011120.0001 BACE4011120.0005
4-Fluoro-D-phenylalanine
4-Fluoro-D-phenylalanine
Formule: C₉H₁₀FNO₂
MDL Nummer: MFCD00063063
CAS nummer: 18125-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR