2,4-Dimethylpyrrole ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,4-Dimethyl-1H-pyrrole
D2848-1G
D2848-5G
D2848-25G
TCIAD2848-1GEA
28.6
EUR
TCIAD2848-1G
TCIAD2848-5G
TCIAD2848-25G
2,4-Dimethylpyrrole ≥97.0% (by GC)
2,4-Dimethylpyrrole
Formule:
C₆H₉N MW: 95.1424 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00192088 CAS nummer: 625-82-1 EINECS: 210-912-2 |
|
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...