Monomethyl terephthalate ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T0244-25G T0244-500G
TCIAT0244-25GEA 46.2 EUR
TCIAT0244-25G TCIAT0244-500G
Monomethyl terephthalate ≥98.0% (by GC, titration analysis)
Monomethyl terephthalate
Formule: C₉H₈O₄
MDL Nummer: MFCD00002557
CAS nummer: 1679-64-7
EINECS: 216-849-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f40e93879d79be5b0c6ee5846e1cd64751b59e9e&vid=f40e93879d79be5b0c6ee5846e1cd64751b59e9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 221.0 to 224.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR