2,5-Dihydroxyterephthalic acid ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D3899-25G
D3899-5G
TCIAD3899-25GEA
280
EUR
TCIAD3899-25G
TCIAD3899-5G
2,5-Dihydroxyterephthalic acid ≥98.0% (by HPLC, titration analysis)
2,5-Dihydroxyterephthalic acid
Formule:
C₈H₆O₆ Smeltpunt: 339°C |
MDL Nummer:
MFCD00132933 CAS nummer: 610-92-4 EINECS: 210-239-4 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...