Methyl cyclohexanecarboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0471-25ML C0471-500ML C0471-100ML
TCIAC0471-25MLEA 17.6 EUR
TCIAC0471-25ML TCIAC0471-500ML TCIAC0471-100ML
Methyl cyclohexanecarboxylate ≥98.0% (by GC)
Methyl cyclohexanecarboxylate
Formule: C₈H₁₄O₂
Vlampunt: 60°C
MDL Nummer: MFCD00001458
CAS nummer: 4630-82-4
EINECS: 225-050-2
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc2d83c490751ab615ed04abf90c54bfc4e61dd&vid=9cc2d83c490751ab615ed04abf90c54bfc4e61dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR