Methyl cyclopentanecarboxylate 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L00741.06
L00741.14
L00741.06EA
72.4
EUR
L00741.06
L00741.14
Methyl cyclopentanecarboxylate 97%
Methyl cyclopentanecarboxylate
Purity: 98%
CAS No.: 4630-80-2
Molecular Formula: C7H12O2
Formula Weight: 128.17
Refractive Index: 1.4360
Boiling Point: 81-82°/5mm
MDL No.: MFCD00019290
Formule:
C₇H₁₂O₂ Kookpunt: 81…82 °C (5 mmHg) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00019290 CAS nummer: 4630-80-2 EINECS: 225-049-7 |