2-Bromo-5-chloro-3-methylbenzo[b]thiophene
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KM08172DA KM08172EA
MAYBKM08172DAEA 0 EUR
MAYBKM08172DA MAYBKM08172EA
2-Bromo-5-chloro-3-methylbenzo[b]thiophene
2-Bromo-5-chloro-3-methylbenzo[b]thiophene
Formule: C₉H₆BrClS
MW: 261.57 g/mol
MDL Nummer: MFCD00126363
CAS nummer: 175203-60-8