Etidronic acid ∼60% ∼4.2 mol/L in water
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
HEDP, OEDFK, 1-Hydroxyethylidenediphosphonic acid, 1-Hydroxyethane-1,1-diphosphonic Acid, HDEPA, 1-Hydroxyethylidene-1,1-diphosphonic acid, 1-Hydroxyethylidenebis(phosphonic acid)
H0587-25G
H0587-500G
TCIAH0587-25GEA
18.7
EUR
TCIAH0587-25G
TCIAH0587-500G
Etidronic acid ∼60% ∼4.2 mol/L in water
Etidronic acid
Formule:
C₂H₈O₇P₂ MW: 206.028 g/mol |
MDL Nummer:
MFCD00070585 CAS nummer: 2809-21-4 EINECS: 220-552-8 UN: 3265 ADR: 8 III Merck Index: 14,03863 |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Concentration (Neutralization titration) | 59.0 to 65.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...