N-Fmoc-D-leucine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4005951.0005 4005951.0025 4005951.0100
BACE4005951.0005EA 0 EUR
BACE4005951.0005 BACE4005951.0025 BACE4005951.0100
N-Fmoc-D-leucine
N-Fmoc-D-leucine
Formule: C₂₁H₂₃NO₄
MDL Nummer: MFCD00062957
CAS nummer: 114360-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR