N-Fmoc-L-leucine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NRB05198.1 NRB05198EA NRB05198EE
MAYBNRB05198.1EA 0 EUR
MAYBNRB05198.1 MAYBNRB05198EA MAYBNRB05198EE
N-Fmoc-L-leucine
N-Fmoc-L-leucine
Formule: C₂₁H₂₃NO₄
MDL Nummer: MFCD00037133
CAS nummer: 35661-60-0
EINECS: 252-662-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR