(R)-(+)-α-Lipoic acid, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
04471-100MGEA
133
EUR
04471-100MG
(R)-(+)-α-Lipoic acid, Supelco®
(R)-(+)-α-Lipoic acid
Formule:
C₈H₁₄O₂S₂ MW: 206,3256 g/mol |
MDL Nummer:
MFCD01631142 CAS nummer: 1200-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...