N-Fmoc-L-leucine-[¹⁵N]

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4026149.0100 4026149.0250 4026149.1000
BACE4026149.0100EA 0 EUR
BACE4026149.0100 BACE4026149.0250 BACE4026149.1000
N-Fmoc-L-leucine-[¹⁵N]
N-Fmoc-L-leucine-[¹⁵N]
Formule: C₂₁H₂₃NO₄
MDL Nummer: MFCD00209748
CAS nummer: 200937-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR