Ultracel YM Ultrafiltration Membrane Disc Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13651 14412 14422AM 14432 14442AM 14451 14461
MILF13651EA 0 EUR
MILF13651 MILF14412 MILF14422AM MILF14432 MILF14442AM MILF14451 MILF14461
Ultracel YM Ultrafiltration Membrane Disc Filters
Filters Membraanfilters
Ultrafiltration cut disc membranes are available in Ultracel® regenerated cellulose and Biomax® polyethersulvone (PES).
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic and tight microstructure assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.