Laboratory chairs, standing rests, stools, Labster
Leverancier: BIMOS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
132-0481EA 490 EUR
132-0481 132-0482 132-0460 132-0480 132-0463 132-0464 132-0461 132-0462 132-0478 132-0456 132-0973 132-0479 132-0457 132-0974 132-0971 132-0476 132-0454 132-0477 132-0972 132-0455 132-0458 132-0459 132-0470 132-0471 132-0452 132-0474 132-0453 132-0970 132-0475 132-0472 132-0451 132-0473 139-0758 139-0759 BIMO9107-6902-807 132-0968 132-0969 139-0761 132-0467 132-0468 139-0461 132-0465 132-0466 139-0760 132-0469
Laboratory chairs, standing rests, stools, Labster
Stoelen
Apart from special hygiene features, a laboratory chair must always be ready for use at changing places of work. It should take up a minimum of space yet still offer maximum ergonomics and seating comfort so as to guarantee precise and concentrated work over longer periods of time. Labster is ideal for everyday use in the laboratory.
  • Specifically designed for laboratory work, with no sharp edges
  • The rounded surface geometries ensures clean lines, without crevices to catch dirt, allowing quick and thorough cleaning
Labster 2 and Labster 3 chairs
The Auto-Motion technology supports the bent over posture
Seat and backrest height adjustment
Ergonomically shaped seat and backrest upholstery
Plastic star base improves stability, flat and compact design like an open umbrella to allow optimum cleaning, the design also ensures that the base is not a stumbling block in typically cramped laboratory working environment
Twin castors for hard floors for Labster 2 ; floor glides and height adjustable plastic foot ring for Labster 3
Plastic parts in platinum grey
Chrome plated steel parts
Easy cleaning thanks to smooth surfaces
Labster standing rests
The Labster standing aid is the solution for workplaces where the user can only work standing up. It allows dynamic 'upright sitting' and takes the stain off the user‘s spine. Its pendulum base follows the user's movements during work and allows him to reach out on all sides without having to stand up. The Labster stool takes up hardly any space under the desk , when not in use. Everything fits with this system: for example, the round base of the stool into the star base of the chair. So that two specialists share the space of one.
Ideal for short periods of sitting.
Its plastic round base means it can‘t get caught anywhere and makes the stool very mobile
The 7° pendulum base of the standing aid ensures movement
No nooks or crannies where micro-organisms, bacteria or minute particles can collect, all joins between parts are sealed
Seat height adjustment with ring release
Seat with anti-slip properties and small backrest
Large working radius
Plastic parts in platinum grey
Chrome plated steel parts
Easy cleaning thanks to smooth surfaces
Labster stools
Seat height adjustment with ring release
Provide ad hoc seating and can be hidden underneath the workbench
Disc base with no corners or edges (no 5 star base so there is no risk of getting caught)
Twin castors for hard floors
Compact and space saving
Completely washable; all materials are resistant to disinfectants, seamless
Certificaten: Suitable for all laboratory and hygiene-critical areas
GMP compatible
Suitable for cleanrooms and class 3 in air purity acc. to DIN EN ISO 14644-1 and air purity class 1 acc. to US-Fed St. 209 E
Suitable for bio-laboratories in safety classes S1, S2 and S3
GS symbol for tested safety
Corresponds to DIN 68877
Fraunhofer Tested Device