Gas washing bottles, Drechsel pattern, with head
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41469012 41469018 41469024 41469030
HECH41469012EA 64.7 EUR
HECH41469012 HECH41469018 HECH41469024 HECH41469030
Gas washing bottles, Drechsel pattern, with head
Flessen Gaswasflessen
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Without filter disc.
Certificaten: DIN 12596