Cap mats and strips
Leverancier: THERMO MATRIX TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4411 4412 4413 4414 4415 4416 4421 4422 4410 4430 4431 4432
MATR4411EA 0 EUR
MATR4411 MATR4412 MATR4413 MATR4414 MATR4415 MATR4416 MATR4421 MATR4422 MATR4410 MATR4430 MATR4431 MATR4432
Cap mats and strips
Afdichtingen Afdichtingen van microtiterplaten
Matrix CapMats and CapStrips are engineered to maintain a strong seal at all times on tubes, microplates, and deepwell blocks. CapMats effectively seal down to –80 °C, while cap strips give the flexibility to seal individual caps or rows.
  • Low temperature compatible so seals remain strong even under extreme conditions
  • Securely seals plates, blocks and tubes ensures sample integrity no matter what the vessel
  • EVA material construction provides a chemically resistant seal
  • Alphanumeric designation on mats simplifies well identification
Cap mats are theMore Product Information