Screw top vials and closures, 13 mm, CrossLab
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8010-0021 8010-0556 8010-0555 8010-0554 8010-0553 8010-0559 8010-0562 8010-0561 8010-0560
HEWL8010-0021EA 0 EUR
HEWL8010-0021 HEWL8010-0556 HEWL8010-0555 HEWL8010-0554 HEWL8010-0553 HEWL8010-0559 HEWL8010-0562 HEWL8010-0561 HEWL8010-0560
Screw top vials and closures, 13 mm, CrossLab
flesjes
These screw top vials have precision-formed screw threads 13-425 for consistent secure seal and can be used as wash, waste, or storage vials.
  • Compatible with WISP 48 style autosamplers