PIG® clear compact spill kit
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KITL250 KITL350 KITL450
121-0075EA 46.8 EUR
121-0075 121-0076 121-0077
PIG® clear compact spill kit
Absorptiematerialen en bescherming tegen morsen
See-through, compact spill response Kit makes it easy to know when to replenish supplies.
  • Hook and loop closure is easy to open for fast access to absorbents
  • Carry handle affords convenient transport makes it ideal for hanging on a wall, storing in a cabinet or being carried in a vehicle cab
  • Compact design takes up little storage room and effectively deals with minor spillages
  • Includes a selection of absorbents such as PIG® Mats to absorb the contained liquid and a PIG® Pillow for larger spills or persistent drips
Contains temporary disposal bags and ties for neat and convenient disposal of used absorbents.
Verpakking: In a durable see-through wallet that makes it simple to check spill response supplies with a quick glance.
Opgelet: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.