Snap top vials and closures, 11 mm, CrossLab
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8010-0024 8010-0026 8010-0177 8010-0178 8010-0411 8010-0200
HEWL8010-0024EA 0 EUR
HEWL8010-0024 HEWL8010-0026 HEWL8010-0177 HEWL8010-0178 HEWL8010-0411 HEWL8010-0200
Snap top vials and closures, 11 mm, CrossLab
flesjes
These snap top vials made of glass with precision-formed neck improves autosampler handling.
  • Compatible with 11 mm snap or crimp cap
  • Optional ceramic write-on spot
  • Available in unique packaging designed to reduce vial breakage