Top tilt measures
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41877006 41877018 41877009 41877012 41877003 41877015
HECH41877006EA 30.1 EUR
HECH41877006 HECH41877018 HECH41877009 HECH41877012 HECH41877003 HECH41877015
Top tilt measures
Flesdispenseropzet
Automatic tilt measures, made of DURAN® borosilicate glass 3.3, with standard ground cone NS 29/32. Supplied without reservoir bottle.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes