TSK-Gel® alpha and superaw columns for gel filtration
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0018340 0018341 0018345 0019315 0019316 0019317 0019318 0019319 0019320 0018343 0019321 0019322 0018344 0018342 0020011 0020012 0018339
TOYO0018340EA 2210 EUR
TOYO0018340 TOYO0018341 TOYO0018345 TOYO0019315 TOYO0019316 TOYO0019317 TOYO0019318 TOYO0019319 TOYO0019320 TOYO0018343 TOYO0019321 TOYO0019322 TOYO0018344 TOYO0018342 TOYO0020011 TOYO0020012 TOYO0018339
TSK-Gel® alpha and superaw columns for gel filtration
Kolommen voor chromatografie
These columns are packed with a hydrophilic, highly crosslinked polymer used for a wide range of solvents. Suitable for the separations of synthetic polymers, oligomers, additives and detergents, saccharides, nucleic acids and peptides.
  • Unique hydrophilic polymethacrylate resin with rigid spherical beads
  • Minimal swelling characteristics from 100% water to 100% non-polar solvents
  • Excellent mechanical and chemical stability
  • High resolution power and short analysis time