Membrane filters, PORAFIL® CA
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
68000080050 68000020013 68000020025 68000020047 68000020050 68000020090 68000020100 68000020142 68000020220 68000045013 68000045025 68000045047 68000045050 68000045090 68000045100 68000045142 68000045220 68000080013 68000080025 68000080047 68000080090 68000080100 68000080142 68000080220
MANA68000080050EA 192 EUR
MANA68000080050 MANA68000020013 MANA68000020025 MANA68000020047 MANA68000020050 MANA68000020090 MANA68000020100 MANA68000020142 MANA68000020220 MANA68000045013 MANA68000045025 MANA68000045047 MANA68000045050 MANA68000045090 MANA68000045100 MANA68000045142 MANA68000045220 MANA68000080013 MANA68000080025 MANA68000080047 MANA68000080090 MANA68000080100 MANA68000080142 MANA68000080220
Membrane filters, PORAFIL® CA
Filters Membraanfilters
Cellulose acetate membranes have a low protein binding capacity and are suited for aqueous and alcoholic media.
  • Hydrophilic
  • Suitable for use with hot gases up to 180 °C
  • Sterilisable