Disruptor Beads™ for Disruptor Genie®
Leverancier: SCIENTIFIC INDUSTRIES ELMIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SI-BG01 SI-BG05
432-0051EA 77.9 EUR
432-0051 432-0052
Disruptor Beads™ for Disruptor Genie®
Parels Glasparels
Glass beads are designed for use with the Disruptor Genie™ or TurboMix™ attachment for optimum disruption of bacteria, yeast, fungi and other cells.
Two sizes are available, 0,1 mm (bacteria), 0,5 mm (yeast, fungi and other sample types).