Sample Loops for Rheodyne® Valves
Leverancier: Agilent Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HEWL0101-1229EA 168 EUR
HEWL0101-1229 HEWL0101-1228 HEWL1535-4860 HEWL0101-1234 HEWL0101-1236 HEWL0101-1235 HEWL0101-1238 HEWL0101-1237 HEWL0101-1250 HEWL0101-1252 HEWL0101-1230 HEWL0101-1251 HEWL0101-1239 HEWL0101-1219 HEWL0101-1243 HEWL0101-1242 HEWL0100-1924 HEWL0101-0376 HEWL0101-1245 HEWL0101-1244 HEWL0100-1921 HEWL0101-0378 HEWL0101-1247 HEWL0101-0377 HEWL0101-1246 HEWL0101-1227 HEWL0101-1249 HEWL0100-1923 HEWL0101-0379 HEWL0101-1248 HEWL0101-1226 HEWL0100-1922 HEWL0101-1241 HEWL0101-1240
Sample Loops for Rheodyne® Valves
Benodigdheden voor chromatografie Injectieloops voor chromatografiesystemen
These PEEK sample loops are inert to most organic solvents. The stainless-steel sample loops are not interchangeable due to the change in port angle.
  • Stainless steel loops are square cut and free of burrs for a flush connection
  • Flexible PEEK loops have a clean, straight cut for low dead volume connections
Accuracy of large PEEK loops is ±14%, intermediate loops ±21%, small loops ±65%. Accuracy of large metal loops is ±5%, intermediate loops ±10%, small loops ±30%