Gooch crucibles
Leverancier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L82A/2 L82A/3 L82R/3
HALDL82A/2EA 24.2 EUR
HALDL82A/2 HALDL82A/3 HALDL82R/3
Gooch crucibles
Kroezen
Porcelain gooch crucibles, with perforated base, glazed.
  • For use in temperatures up to 1150 °C
Ideal for total suspended solids determination.