Crucibles, Alsint 99,7, high form
Leverancier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LALC/00 LALC/0 LALC/10 LALC/1 LALC/2 LALC/3 LALC/4 LALC/5 LALC/6 LALC/7 LALC/8 LALC/9
459-1611EA 30.1 EUR
459-1611 459-1612 459-1622 459-1613 459-1614 459-1615 459-1616 459-1617 459-1618 459-1619 459-1620 459-1621
Crucibles, Alsint 99,7, high form
Kroezen
These crucibles and lids are made of Alsint 99,7.
  • Withstand maximum temperature of 1700 °C
  • Suitable for crystal growing, fusion processes, annealing
Certificaten: According to DIN EN 60672, Group C700, Type C799