GLP-kits, Finnpipette™ Novus
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4701120 4701130 4700470 4700490
613-1428EA 1309 EUR
613-1428 613-1429 613-5588 613-5589
GLP-kits, Finnpipette™ Novus
Pipetten Micropipetten
Access everything needed for pipetting in Finnpipette™ Novus GLP kits. These all-inclusive kits feature ergonomic and easy to use Finnpipette™ Novus pipetters. The kits contains four different volume range to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
  • Greater ease-of-use and reduced risk of injury
  • Adjustable index-finger trigger action prevents common repetitive stress injuries caused by thumb-driven pipettes
  • Easy-to-follow prompts guide Novus users through 10 pipetting functions
Alle kits bevatten eenMore Product Information