Strips of 8 PCR tubes with attached caps
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
608281 608283_120 608284_120 608286_120 608201_125 608281UK
GREI608281EA 0 EUR
GREI608281 GREI608283_120 GREI608284_120 GREI608286_120 GREI608201_125 GREI608281UK
Strips of 8 PCR tubes with attached caps
Buisjes PCR-buisjes
PP, natural.
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Free of heavy metals
  • Temperature range: –20 °C to +105 °C
Comprehensive thermal cycler compatibility.
Verpakking: Packaging labelled with product symbols for easier identification and warehousing.