TSKgel® universal reversed phase columns
Leverancier: Tosoh Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TSK-Gel®
TOYO0022756EA 563 EUR
TOYO0022756 TOYO0022712 TOYO0022711 TOYO0022755 TOYO0022754 TOYO0021940 TOYO0022710 TOYO0022753 TOYO0022752 TOYO0022751 TOYO0022750 TOYO0021462 TOYO0021829 TOYO0022719 TOYO0022718 TOYO0022717 TOYO0022716 TOYO0022715 TOYO0022759 TOYO0022758 TOYO0022714 TOYO0022757 TOYO0022713 TOYO0022745 TOYO0022701 TOYO0021458 TOYO0021810 TOYO0022700 TOYO0021457 TOYO0022744 TOYO0022743 TOYO0021456 TOYO0022742 TOYO0021455 TOYO0021454 TOYO0022741 TOYO0021453 TOYO0022740 TOYO0022709 TOYO0021939 TOYO0021938 TOYO0022708 TOYO0022707 TOYO0022706 TOYO0022749 TOYO0022705 TOYO0021814 TOYO0022748 TOYO0022704 TOYO0021813 TOYO0022747 TOYO0021812 TOYO0022703 TOYO0022746 TOYO0021811 TOYO0022702 TOYO0021459 TOYO0021461 TOYO0021460 TOYO0021843 TOYO0022734 TOYO0022733 TOYO0021842 TOYO0021841 TOYO0022732 TOYO0022731 TOYO0022730 TOYO0022739 TOYO0022738 TOYO0022737 TOYO0022736 TOYO0022735 TOYO0021844 TOYO0022723 TOYO0021997 TOYO0022722 TOYO0021996 TOYO0021831 TOYO0022765 TOYO0022721 TOYO0021830 TOYO0021995 TOYO0022720 TOYO0022764 TOYO0022763 TOYO0022762 TOYO0022761 TOYO0022760 TOYO0021839 TOYO0022729 TOYO0021838 TOYO0022728 TOYO0022727 TOYO0022726 TOYO0022725 TOYO0022724
TSKgel® universal reversed phase columns
Kolommen voor chromatografie
TSKgel ODS-100V/ODS-100Z columns incorporate the best-inclass surface properties to limit secondary interactions of basic, acidic and chelating compounds.
  • The ultra purity Type B base silica contains negligible amounts of metal ion impurities, minimizes sample adsorption
  • High surface area (450 m²/g) silica
  • Spherical 3 and 5 µm particles with 10 nm (100 Å) pores
  • Very high column efficiency and moderate column back pressure
  • Monomeric bonding chemistry
  • Low residual silanol content
TSKgel ODS-100V provides strong retention for polar compounds due to its lower C18 ligand density (15% carbon content). Proprietary monomeric bonded phase chemistry provides complete wetting and retention stability in 100% aqueous mobile phases.
TSKgel ODS-100Z contains a high density (20% carbon content) monomeric C18 bonded phase for maximum retention and selectivity of small molecular weight compounds. Exhaustive endcapping prevents secondary interaction with residual silanol groups.
Both column lines were expanded to include 3 µm packed columns. These columns are well suited for high throughput LC/MS applications, providing fast and efficient separations.
These columns used for analysis of water and lipid soluble vitamins, organic acids, polymer additives and nucleotides (mono, di and tri).