Capillary electrochromatography (CEC) capillaries
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5063-6512 5063-6513 5063-6535 5063-6536 5063-6540 5063-6541
HEWL5063-6512EA 0 EUR
HEWL5063-6512 HEWL5063-6513 HEWL5063-6535 HEWL5063-6536 HEWL5063-6540 HEWL5063-6541
Capillary electrochromatography (CEC) capillaries
Elektroforesesystemen Systemen voor capillaire elektroforese
Capillary electrochromatography is a hybrid of CE and LC and can be performed in the Agilent CE system. Using CE capillaries packed with LC stationary phases, CEC offers the loadability and selectivity of LC and the high efficiency of CE.
Using the high pressure capabilities of the Agilent CE system, both ends of the CEC capillary can be pressurised. This process prevents outgassing upon application of high voltage and significantly extends capillary lifetime.
Use CEC to improve resolution of solutes, which are difficult to resolve by HPLC, for hydrophobic solutes which cannot be solubilised in MEKC buffers, or for reduced sample and solvent consumption compared to HPLC.
The colour coding of the capillary (alignment stopper) and the interface makes it easy to combine the correct alignment interface with the capillary.