Cross-linked and bonded μSIL capillaries
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
194-8111 196-7203 199-2602 196-1002
HEWL194-8111EA 432 EUR
HEWL194-8111 HEWL196-7203 HEWL199-2602 VARI196-1002
Cross-linked and bonded μSIL capillaries
Elektroforesesystemen Systemen voor capillaire elektroforese
A series of coated capillaries specifically designed for CE, which are prepared by cross-linking and bonding a novel, proprietary fluorocarbon (FC) polymer. μSIL-FC capillaries are chemically inert, hydrophobic, and stable from pH 2,5 to 10.
These capillaries are a must-have for cIEF, protein, peptide and carbohydrate separations, as well as replaceable gel CE applications such as oligonucleotides, DNA fragments, and PCR product separations.
μSIL-DNA capillaries are also coated with an FC polymer but have a 75 μm ID to accommodate the viscosity of entangled polymer solutions. All μSILMore Product Information